EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Muscarinic M2 antagonists: anthranilamide derivatives with exceptional selectivity and in vivo activity.
PMID 14723952
Abstract
Anthranilamide analogues such as 23 are potent and highly selective muscarinic M2 antagonists that also show good oral bioavailability and in vivo activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
461350
1-Boc-4-piperidone, 98%
C10H17NO3