EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Microwave assisted Leimgruber-Batcho reaction for the preparation of indoles, azaindoles and pyrroylquinolines.
PMID 14737637
Abstract
The development of enhanced conditions for Lewis acid catalysed Leimgruber-Batcho indole synthesis using microwave acceleration is described. This approach has permitted the preparation of a variety of heteroaromatic enamine intermediates in good yield and high purities. Subsequent catalytic hydrogenation reactions, under various conditions including the use of a solid-phase encapsulated catalyst, furnish the corresponding indole derivatives in good yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
684910
1H-Benzo[g]indole, 97%
C12H9N
685488
1H-Pyrrolo[2,3-f]quinoline, 97%
C11H8N2