EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Nuclear envelope assembly following mitosis.
PMID 1488014
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62962 L-α-Lysophosphatidylcholine, from egg yolk, ≥99.0% (TLC)