EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Biochemical characterization of the mitochondrial tRNASer(UCN) T7511C mutation associated with nonsyndromic deafness.
PMID 14960712
Abstract
We report here the biochemical characterization of the deafness-associated mitochondrial tRNA(Ser(UCN)) T7511C mutation, in conjunction with homoplasmic ND1 T3308C and tRNA(Ala) T5655C mutations using cybrids constructed by transferring mitochondria from lymphoblastoid cell lines derived from an African family into human mtDNA-less (rho degrees ) cells. Three cybrids derived from an affected matrilineal relative carrying the homoplasmic T7511C mutation, exhibited approximately 75% decrease in the tRNA(Ser(UCN)) level, compared with three control cybrids. This amount of reduction in the tRNA(Ser(UCN)) level is below a proposed threshold to support a normal rate of mitochondrial protein synthesis in lymphoblastoid cell lines. This defect is likely a primary contributor to approximately 52% reduction in the rate of mitochondrial protein synthesis and marked defects in respiration and growth properties in galactose-containing medium. Interestingly, the T5655C mutation produces approximately 50% reduction in the tRNA(Ala) level in mutant cells. Strikingly, the T3308C mutation causes a significant decrease both in the amount of ND1 mRNA and co-transcribed tRNA(Leu(UUR)) in mutant cells. Thus, mitochondrial dysfunctions caused by the T5655C and T3308C mutations may modulate the phenotypic manifestation of the T7511C mutation. These observations imply that a combination of the T7511C mutation with two mtDNA mutations accounts for the high penetrance of deafness in this family.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03353583910
DIG Oligonucleotide Tailing Kit, 2nd generation