EMAIL THIS PAGE TO A FRIEND
RNA (New York, N.Y.)
Functional group recognition at the aminoacylation and editing sites of E. coli valyl-tRNA synthetase.
PMID 14970394
Abstract
To correct misactivation and misacylation errors, Escherichia coli valyl-tRNA synthetase (ValRS) catalyzes a tRNA(Val)-dependent editing reaction at a site distinct from its aminoacylation site. Here we examined the effects of replacing the conserved 3'-adenosine of tRNA(Val) with nucleoside analogs, to identify structural elements of the 3'-terminal nucleoside necessary for tRNA function at the aminoacylation and editing sites of ValRS. The results show that the exocyclic amino group (N6) is not essential: purine riboside-substituted tRNA(Val) is active in aminoacylation and in stimulating editing. Presence of an O6 substituent (guanosine, inosine, xanthosine) interferes with aminoacylation as well as posttransfer and total editing (pre- plus posttransfer editing). Because ValRS does not recognize substituents at the 6-position, these results suggest that an unprotonated N1, capable of acting as an H-bond acceptor, is an essential determinant for both the aminoacylation and editing reactions. Substituents at the 2-position of the purine ring, either a 2-amino group (2-aminopurine, 2,6-diaminopurine, guanosine, and 7-deazaguanosine) or a 2-keto group (xanthosine, isoguanosine), strongly inhibit both aminoacylation and editing. Although aminoacylation by ValRS is at the 2'-OH, substitution of the 3'-terminal adenosine of tRNA(Val) with 3'-deoxyadenosine reduces the efficiency of valine acceptance and of posttransfer editing, demonstrating that the 3'-terminal hydroxyl group contributes to tRNA recognition at both the aminoacylation and editing sites. Our results show a strong correlation between the amino acid accepting activity of tRNA and its ability to stimulate editing, suggesting misacylated tRNA is a transient intermediate in the editing reaction, and editing by ValRS requires a posttransfer step.
Related Materials