EMAIL THIS PAGE TO A FRIEND
FEBS letters
Detergents as tools for the purification and classification of lipid rafts.
PMID 14986659
Abstract
The relative insolubility of lipid rafts in cold non-ionic detergents is the most widely used method to purify these fascinating membrane domains from intact cells or membranes. Most of what we know about lipid raft function has been derived from experiments utilising detergent insolubility as the basis for raft purification. Recently, a wider range of detergents have been used to purify 'rafts', and rafts have been subclassified based on their differential solubility in different detergents. This minireview critically examines the use of detergents as tools for raft isolation and for the subclassification of rafts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X100RS
Triton X-100 reduced