EMAIL THIS PAGE TO A FRIEND
BioTechniques
Preparation of genomic DNA from Dictyostelium discoideum for PCR analysis.
PMID 15088372
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8312
REDTaq® Genomic DNA Polymerase, with MgCl2