EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The RGS (regulator of G-protein signalling) and GoLoco domains of RGS14 co-operate to regulate Gi-mediated signalling.
PMID 15112653
Abstract
RGS (regulator of G-protein signalling) proteins stimulate the intrinsic GTPase activity of the a subunits of heterotrimeric G-proteins, and thereby negatively regulate G-protein-coupled receptor signalling. RGS14 has been shown previously to stimulate the GTPase activities of Ga(o) and Ga(i) subunits through its N-terminal RGS domain, and to down-modulate signalling from receptors coupled to G(i). It also contains a central domain that binds active Rap proteins, as well as a C-terminal GoLoco/G-protein regulatory motif that has been shown to act in vitro as a GDP-dissociation inhibitor for Ga(i). In order to elucidate the respective contributions of the three functional domains of RGS14 to its ability to regulate G(i) signalling, we generated RGS14 mutants invalidated in each of its domains, as well as truncated molecules, and assessed their effects on G(i) signalling via the bg pathway in a stable cell line ectopically expressing the G(i)-coupled M2 muscarinic acetylcholine receptor (HEK-m2). We show that the RGS and GoLoco domains of RGS14 are independently able to inhibit signalling downstream of G(i). Targeting of the isolated GoLoco domain to membranes, by myristoylation/palmitoylation or Rap binding, enhances its inhibitory activity on G(i) signalling. Finally, in the context of the full RGS14 molecule, the RGS and GoLoco domains co-operate to confer maximal activity on RGS14. We therefore propose that RGS14 combines the inhibition of G(i) activation or coupling to receptors via its GoLoco domain with stimulation of the GTPase activity of Ga(i)-GTP via its RGS domain to negatively regulate signalling downstream of G(i).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3884 EclX I from Enterobacter cloacae 590, Restriction Enzyme