EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A mild method for the preparation of gamma-hydroxy-alpha,beta-acetylenic esters.
PMID 15127442
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
274879
1,4-Cyclohexanedione monoethylene acetal, 97%
C8H12O3