EMAIL THIS PAGE TO A FRIEND
Cancer research
A novel role for mixed-lineage kinase-like mitogen-activated protein triple kinase alpha in neoplastic cell transformation and tumor development.
PMID 15172994
Abstract
Previously, no member of the mixed-lineage kinase (MLK) protein family was known to function as an oncogene. Here, we demonstrate that MLK-like mitogen-activated protein triple kinase (MLTK)-alpha, a member of the MLK family, induced neoplastic cell transformation and tumorigenesis in athymic nude mice. Introduction of small interference RNA (siRNA)-MLTK-alpha into MLTK-alpha-overexpressing cells dramatically suppressed cell transformation. Nuclear accumulation of the pHisG-MLTK-alpha fusion protein was observed after epidermal growth factor or 12-O-tetradecanoylphorbol-13-acetate treatment. Phosphorylation of downstream mitogen-activated protein kinase-targeted transcription factors including c-Myc, Elk-1, c-Jun, and activating transcription factor (ATF) 2 was also differentially enhanced in MLTK-alpha-overexpressing cells exposed to epidermal growth factor or 12-O-tetradecanoylphorbol-13-acetate stimulation compared with cells expressing mock vector or siRNA-MLTK-alpha. Very importantly, MLTK-alpha-overexpressing cells formed fibrosarcomas when injected s.c. into athymic nude mice, whereas almost no tumor formation was observed in mice that received injections of mock or siRNA-MLTK-alpha stably transfected cells. These results are the first to indicate that MLTK-alpha plays a key role in neoplastic cell transformation and cancer development.