EMAIL THIS PAGE TO A FRIEND
Organic letters
Expedient protocol for solid-phase synthesis of secondary and tertiary amines.
PMID 15176787
Abstract
[reaction: see text] An expedient solid-phase synthetic approach to secondary and tertiary amines was developed. The protocol employs conversion of resin-bound amino alcohols to the corresponding iodides, followed by iodide displacement with primary or secondary amines or with unprotected amino alcohols. This two-step procedure, affording products in good to excellent yields, is suitable for solid-phase synthesis of polyamines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
497312
4-Piperidinemethanol, 97%
C6H13NO