EMAIL THIS PAGE TO A FRIEND
Genome biology
POSaM: a fast, flexible, open-source, inkjet oligonucleotide synthesizer and microarrayer.
PMID 15287980
Abstract
DNA arrays are valuable tools in molecular biology laboratories. Their rapid acceptance was aided by the release of plans for a pin-spotting microarrayer by researchers at Stanford. Inkjet microarraying is a flexible, complementary technique that allows the synthesis of arrays of any oligonucleotide sequences de novo. We describe here an open-source inkjet arrayer capable of rapidly producing sets of unique 9,800-feature arrays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
65290
3-Methoxypropionitrile, ≥98.0% (GC)
C4H7NO