EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
1,5-(Diethylamino)piperidine, a novel spermidine analogue that more specifically activates the N-methyl-D-aspartate receptor-associated polyamine site.
PMID 1535417
Abstract
We have investigated the action of 1,5-(diethylamino)piperidine (DEAP), a novel spermidine analogue that activates the polyamine site associated with the N-methyl-D-aspartate receptor. DEAP increased [3H]dizocilpine ([3H]MK801) binding to rat brain membranes with a potency similar to that of spermine and spermidine, but with a somewhat greater efficacy. Unlike other polyamines, however, DEAP did not exhibit low affinity inhibition of [3H] dizocilpine binding, suggesting that it binds more selectively to the polyamine site. DEAP increased the equilibrium affinity of [3H]dizocilpine. The increase in affinity was due to slowed dissociation, as well as a small increase in the association rate of [3H]dizocilpine. All of these effects of DEAP could be reversed by arcaine. These data illustrate the utility of DEAP as a novel polyamine agonist at the N-methyl-D-aspartate receptor complex. However, these data fail to support the hypothesis that polyamines activate the N-methyl-D-aspartate receptor by a mechanism similar to that of glutamate and glycine.