EMAIL THIS PAGE TO A FRIEND
Organic letters
Total synthesis of (+)-cocaine via desymmetrization of a meso-dialdehyde.
PMID 15355038
Abstract
[reaction: see text] The total synthesis of (+)-cocaine is described. An extension of the recently reported proline catalyzed intramolecular enol-exo-aldol reaction to a meso-dialdehyde provided the tropane ring skeleton directly with good enantiomeric excess. The meso-dialdehyde was prepared using a 2-azaallyllithium [3 + 2] cycloaddition to generate a cis-2,5-disubstituted pyrrolidine. Overall, the synthesis proceeded in 6.5% yield and 86% ee over 14 linear steps starting from commercially available 3-benzyloxy-1-propanol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
382035
3-Benzyloxy-1-propanol, 97%
C10H14O2