EMAIL THIS PAGE TO A FRIEND
Experimental physiology
Evidence for Na(+)-Ca2+ exchange and Ca(2+)-induced Ca2+ release in a cultured vascular smooth muscle cell line from the rat.
PMID 1543582
Abstract
Measurements of intracellular calcium (Cai2+) and sodium (Nai+) have been made in single smooth muscle cells from the rat aortic cell line (A10) using the Ca(2+)- and Na(+)-sensitive dyes Fura-2 and SBFI (sodium-binding benzofuran isophthalate). The effects of manipulation of intracellular and extracellular Na+ on Cai2+ have been investigated. Reversal of the Na+ gradient in control cells does not result in any measurable increase in Cai2+ or change in the rate of recovery of the cells from agonist stimulation, suggesting that there is little functional Na(+)-Ca2+ exchange. In ouabain-pre-treated cells however, the recovery from agonist stimulation is significantly slowed, suggesting that in the presence of an elevated intracellular Na+ concentration there is an alteration in the Ca(2+)-handling mechanisms. Reversal of the Na+ gradient in ouabain-pre-treated cells results in a transient increase in Cai2+ followed by a slow secondary rise. The transient component of this rise is absent on a second activation of the cell or by prior mobilization of the intracellular stores of Ca2+ by agonist. Data presented in this paper suggest the possibility that the transient component is due to a Ca(2+)-induced Ca(2+)-release mechanism triggered by an initial influx of Ca2+. The mechanism underlying this influx is not known but may involve the Na(+)-Ca2+ exchanger operating in reverse. The possible modulation of the Na(+)-Ca2+ exchanger and Ca(2+)-induced Ca2+ release by internal Na+ is discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93040117
A10-85