EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Crystal structure communications
Hydrogen-bonding patterns in cis-4-ammoniocyclohexanecarboxylate hemihydrate.
PMID 15467151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
404853
cis-4-Aminocyclohexanecarboxylic acid, 95%
C7H13NO2