EMAIL THIS PAGE TO A FRIEND
Nature cell biology
Cytoplasmic ubiquitin ligase KPC regulates proteolysis of p27(Kip1) at G1 phase.
PMID 15531880
Abstract
The cyclin-dependent kinase inhibitor p27(Kip1) is degraded at the G0-G1 transition of the cell cycle by the ubiquitin-proteasome pathway. Although the nuclear ubiquitin ligase (E3) SCF(Skp2) is implicated in p27(Kip1) degradation, proteolysis of p27(Kip1) at the G0-G1 transition proceeds normally in Skp2(-/-) cells. Moreover, p27(Kip1) is exported from the nucleus to the cytoplasm at G0-G1 (refs 9-11). These data suggest the existence of a Skp2-independent pathway for the degradation of p27(Kip1) at G1 phase. We now describe a previously unidentified E3 complex: KPC (Kip1 ubiquitination-promoting complex), consisting of KPC1 and KPC2. KPC1 contains a RING-finger domain, and KPC2 contains a ubiquitin-like domain and two ubiquitin-associated domains. KPC interacts with and ubiquitinates p27(Kip1) and is localized to the cytoplasm. Overexpression of KPC promoted the degradation of p27(Kip1), whereas a dominant-negative mutant of KPC1 delayed p27(Kip1) degradation. The nuclear export of p27(Kip1) by CRM1 seems to be necessary for KPC-mediated proteolysis. Depletion of KPC1 by RNA interference also inhibited p27(Kip1) degradation. KPC thus probably controls degradation of p27(Kip1) in G1 phase after export of the latter from the nucleus.