EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Total synthesis of apoptolidinone.
PMID 15593157
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
663212
Isopropenylboronic acid pinacol ester, contains phenothiazine as stabilizer, 95%
C9H17BO2