EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Role of leucine 31 of phospholamban in structural and functional interactions with the Ca2+ pump of cardiac sarcoplasmic reticulum.
PMID 15644311
Abstract
The ability of two loss-of-function mutants, L31A and L31C, of phospholamban (PLB) to bind to and inhibit the Ca(2+) pump of cardiac sarcoplasmic reticulum (SERCA2a) was investigated using a molecular cross-linking approach. Leu(31) of PLB, located at the cytoplasmic membrane boundary, is a critical amino acid shown previously to be essential for Ca(2+)-ATPase inhibition. We observed that L31A or L31C mutations of PLB prevented the inhibition of Ca(2+)-ATPase activity and disabled the cross-linking of N27C and N30C of PLB to Lys(328) and Cys(318) of SERCA2a. Although L31C-PLB failed to cross-link to any Cys or Lys residue of wild-type SERCA2a, L31C did cross-link with high efficiency to T317C of SERCA2a with use of the homobifunctional sulfhydryl cross-linking reagent, 1,6-bismaleimidohexane. This places Leu(31) of PLB within 10 angstroms of Thr(317) of SERCA2a in the M4 helix. Thus, contrary to previous suggestions, PLB with loss-of-function mutations at Leu(31) retains the ability to bind to SERCA2a, despite losing inhibitory activity. Cross-linking of L31C-PLB to T317C-SERCA2a occurred only in the absence of Ca(2+) and in the presence of nucleotide and was prevented by thapsigargin and by anti-PLB antibody, demonstrating for a fourth cross-linking pair that PLB interacts near M4 only when the Ca(2+) pump is in the Ca(2+)-free, nucleotide-bound E2 conformation, but not in the E2 state inhibited by thapsigargin. L31I-PLB retained full functional and cross-linking activity, suggesting that a bulky hydrophobic residue at position 31 of PLB is essential for productive interaction with SERCA2a. A model for the three-dimensional structure of the interaction site is proposed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
803235
Sulfo-EMCS (N-(ε-maleimidocaproyloxy) sulfosuccinimide ester)
C14H13O9SN2Na