EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Identification of nonpeptidic small-molecule inhibitors of interleukin-2.
PMID 15686897
Abstract
The identification, design, and synthesis of a series of novel sulfamide- and urea-based small-molecule antagonists of the protein-protein interaction IL-2/IL-2Ralpha are described. Installation of a furan carboxylic acid fragment onto a low-micromolar sulfamide resulted in a 23-fold improvement in activity, providing a sub-micromolar, nonpeptidic IL-2 inhibitor (IC(50)=0.60 microM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
433993
Ethyl 5-(chloromethyl)-2-furancarboxylate, 95%
C8H9ClO3