EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Structure-activity relationships and sub-type selectivity in an oxabicyclic estrogen receptor alpha/beta agonist scaffold.
PMID 15713407
Abstract
An oxabicyclic template for estrogen receptor alpha and beta agonists has been identified which can be tuned to provide moderate levels of selectivity for either receptor sub-type. Structure-activity relationships within this phenol-substituted oxabicyclo[3.3.1]nonene series are described. Select compounds from the present series showed activity in vivo after oral dosing in rodent models of uterine proliferation.