EMAIL THIS PAGE TO A FRIEND
Organic letters
Rapid and efficient synthesis of 1,2,4-oxadiazoles utilizing polymer-supported reagents under microwave heating.
PMID 15727476
Abstract
1,2,4-Oxadiazoles can be rapidly and efficiently synthesized from a variety of readily available carboxylic acids and amidoximes using either method A or method B. The use of commercially available polymer-supported reagents combined with microwave heating resulted in high yields and purities of the product 1,2,4-oxadiazoles in an expeditious manner. [structure: see text]