EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Insulin-like growth factor-1 (IGF1) genotype predicts breast volume after pregnancy and hormonal contraception and is associated with circulating IGF-1 levels: implications for risk of early-onset breast cancer in young women from hereditary breast cancer families.
PMID 15756256
Abstract
BRCA1/2 mutations predispose to early-onset breast cancer, especially after oral contraceptive (OC) use and pregnancy. However, the majority of breast cancers might be due to more prevalent low-penetrance genes, which may also modify the risk in BRCA mutation carriers. The absence of the IGF1 19-repeat allele has been associated with high insulin-like growth factor-1 (IGF-1) levels during OC use. High IGF-1 levels are linked to early-onset breast cancer and larger breast volumes in the general population. The goal of this study was to elucidate the relationships between IGF1 genotype, early-onset breast cancer, breast volume, circulating IGF-1 levels and OC use in a prospective cohort of 258 healthy women < or =40 years old from high-risk breast cancer families. All women completed a questionnaire including information on reproductive factors and OC use. We measured the height, weight, breast volumes and plasma IGF-1 levels. IGF-1 levels were similar among parous and nulliparous women not using OCs. In all, 13% had no IGF1 19-repeat allele. There was an interaction between IGF1 genotype and OC use on IGF-1 levels (P=0.026) in nulliparous women and another interaction between IGF1 genotype and parity on breast volume (P=0.01). Absence of the 19-repeat allele was associated with high IGF-1 levels in nulliparous OC users and with larger breast volumes in parous women and OC users. Incident breast cancers were also more common in women without the 19-repeat allele (log rank P=0.002). Our results suggest that lack of the IGF1 19-repeat allele modifies IGF-1 levels, breast volume and possibly early-onset breast cancer risk after hormone exposure in young high-risk women.