EMAIL THIS PAGE TO A FRIEND
Immunity
Rgs1 and Gnai2 regulate the entrance of B lymphocytes into lymph nodes and B cell motility within lymph node follicles.
PMID 15780991
Abstract
Signaling by G protein-coupled receptors coupled to Galpha(i) assists in triggering lymphocyte movement into and out of lymph nodes. Here, we show that modulating the signaling output from these receptors dramatically alters B cell trafficking. Intravital microscopy of adoptively transferred B cells from wild-type and Rgs1-/- mice revealed that Rgs1-/- B cells stick better to lymph node high endothelial venules, home better to lymph nodes, and move more rapidly within lymph node follicles than do wild-type B cells. In contrast, B cells from Gnai2-/- mice enter lymph nodes poorly and move more slowly than do wild-type B cells. The Gnai2-/- mice often lack multiple peripheral lymph nodes, and their B cells respond poorly to chemokines, indicating that Galpha(i1) and Galpha(i3) poorly compensate for the loss of Galpha(i2). These results demonstrate opposing roles for Rgs1 and Gnai2 in B cell trafficking into and within lymph nodes.