EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
New aminocyclitols as modulators of glucosylceramide metabolism.
PMID 15785807
Abstract
A series of 13 aminocyclitol derivatives belonging to two different families is described. Their configuration is governed by the regio- and stereocontrolled epoxide opening of a suitably protected conduritol-B epoxide. Studies on several glycosyl processing enzymes indicate that some of them are good inhibitors of glucosylceramide hydrolase. A rationale to account for preliminary structure-activity relationships is provided.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74089
1-Amino-1-deoxy-scyllo-inositol, ≥95.0% (TLC)
C6H13NO5