EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Cationic chalcone antibiotics. Design, synthesis, and mechanism of action.
PMID 15801857
Abstract
This paper describes how the introduction of "cationic" aliphatic amino groups in the chalcone scaffold results in potent antibacterial compounds. It is shown that the most favorable position for the aliphatic amino group is the 2-position of the B-ring, in particular in combination with a lipophilic substituent in the 5-position of the B-ring. We demonstrate that the compounds act by unselective disruption of cell membranes. Introduction of an additional aliphatic amino group in the A-ring results in compounds that are selective for bacterial membranes combined with a high antibacterial activity against both Gram-positive and -negative pathogens. The most potent compound in this study (78) has an MIC value of 2 muM against methicillin resistant Staphylococus aureus.