EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and photochemotherapeutic activity of thiopyrano[2,3-e]indol-2-ones.
PMID 15837311
Abstract
A series of derivatives of the new ring system thiopyrano[2,3-e]indol-2-one was prepared with the aim of obtaining new photochemotherapeutic drugs. Biological screenings were performed on this new class of photoactivable drugs and a strong antiproliferative effect was observed upon irradiation with UVA light. The compound bearing a methyl substituent at the pyrrole nitrogen resulted as the most interesting showing IC50 in the nanomolar range.