EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis of tricyclic analogs of stephaoxocanidine and their evaluation as acetylcholinesterase inhibitors.
PMID 15878275
Abstract
The synthesis of simplified analogs of the novel isoquinoline alkaloid stephaoxocanidine, carrying the oxazaphenalene ABC-ring system of the natural product, and their activity as inhibitors of the enzyme acetylcholinesterase, are reported. 5,6-Dimethoxy-7H -8-oxa-1-aza-phenalen-9-one (5) was as active as a Narcissus extract enriched in galantamine.