EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
The next generation of phosphodiesterase inhibitors: structural clues to ligand and substrate selectivity of phosphodiesterases.
PMID 15887951
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9766
Dipyridamole, ≥98% (TLC), powder
C24H40N8O4