EMAIL THIS PAGE TO A FRIEND
Organic letters
Suzuki coupling of aryl chlorides with phenylboronic acid in water, using microwave heating with simultaneous cooling.
PMID 15901144
Abstract
[reaction: see text]. We present here a methodology for the Suzuki coupling of aryl chlorides with phenylboronic acid using Pd/C as a catalyst, water as a solvent, and microwave heating. We show that simultaneous cooling in conjunction with microwave heating prolongs the lifetime of the aryl chloride substrates during the course of the reaction and, as a result, yields of the desired biaryl as well as overall recovery of material can be increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78181
Phenylboronic acid, purum, ≥97.0% (HPLC)
C6H7BO2