EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis of substituted indole derivatives as a new class of antimalarial agents.
PMID 15925306
Abstract
A series of substituted indole derivatives were synthesized and evaluated for their in vitro antimalarial activity against P. falciparum. Out of the 24 compounds synthesized six compounds have shown MIC of 1 microg/mL. These compounds are in vitro several folds more active than pyrimethamine.