EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Immobilization of rhodium complexes at thiolate monolayers on gold surfaces: catalytic and structural studies.
PMID 15954778
Abstract
Chiral rhodium-diphosphine complexes have been incorporated into self-assembled thiolate monolayers (SAMs) on gold colloids. Catalysts of this type are of interest because they combine properties of homogeneous and heterogeneous systems. In addition, it should be possible to influence the catalytic properties of the metal center by the neighboring thiolate molecules. Colloids with a diameter of ca. 3 nm, coated with a mixed monolayer of n-octanethiolates and thiolates with chiral rhodium-PYRPHOS end groups, were studied as hydrogenation catalysts. With methyl alpha-acetamido-cinnamate as substrate, virtually the same enantioselectivities (up to 93% ee) and full conversion were obtained as with the corresponding homogeneous [Rh(COD)(PYRPHOS)]BAr(F) catalyst. The colloids were easily recovered by filtration and reused as catalysts three times without loss of enantioselectivity. STM studies of analogous SAMs on Au(111) gave a detailed picture of the structure and dynamics of mixed monolayers of this type. The STM images showed that the catalyst-bearing thiolates are distributed statistically on the surface and that the ordered structure of the n-octanethiolate SAM can be retained during incorporation of the catalyst-bearing thiols using the place-exchange methodology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674397
11-Amino-1-undecanethiol hydrochloride, 99%
C11H25NS · HCl