EMAIL THIS PAGE TO A FRIEND
The FEBS journal
Altered deoxyribonucleotide pools in T-lymphoblastoid cells expressing the multisubstrate nucleoside kinase of Drosophila melanogaster.
PMID 16045762
Abstract
The multisubstrate nucleoside kinase of Drosophila melanogaster (Dm-dNK) can be expressed in human solid tumor cells and its unique enzymatic properties makes this enzyme a suicide gene candidate. In the present study, Dm-dNK was stably expressed in the CCRF-CEM and H9 T-lymphoblastoid cell lines. The expressed enzyme was localized to the cell nucleus and the enzyme retained its activity. The Dm-dNK overexpressing cells showed approximately 200-fold increased sensitivity to the cytostatic activity of several nucleoside analogs, such as the pyrimidine nucleoside analogs (E)-5-(2-bromovinyl)-2'-deoxyuridine (BVDU) and 1-beta-d-arabinofuranosylthymine (araT), but not to the antiherpetic purine nucleoside analogs ganciclovir, acyclovir and penciclovir, which may allow this technology to be applied in donor T cells and/or rescue graft vs. host disease to permit modulation of alloreactivity after transplantation. The most pronounced effect on the steady-state dNTP levels was a two- to 10-fold increased dTTP pool in Dm-dNK expressing cells that were grown in the presence of 1 microm of each natural deoxyribonucleoside. Although the Dm-dNK expressing cells demonstrated dNTP pool imbalances, no mitochondrial DNA deletions or altered mitochondrial DNA levels were detected in the H9 Dm-dNK expressing cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3766
Thymine 1-β-D-arabinofuranoside, ≥99%
C10H14N2O6