EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Asymmetric synthesis of 3,3-diarylpropanals with chiral diene-rhodium catalysts.
PMID 16076189
Abstract
A general route to enantioenriched 3,3-diarylpropanals is presented. These useful building blocks are prepared via an asymmetric rhodium-catalyzed conjugate addition of arylboronic acids to cinnamaldehyde derivatives in the presence of chiral dienes. The addition of both electron-poor as well as electron-rich boronic acids proceeds smoothly with various enals in 63-90% yield with high enantioselectivities (89-93% ee).
Related Materials