EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
ISG15 modification of Ubc13 suppresses its ubiquitin-conjugating activity.
PMID 16112642
Abstract
ISG15 is one of the interferon-stimulated genes and is classified as a ubiquitin-like protein. Upon interferon stimuli, ISG15 is upregulated and becomes conjugated to various cellular proteins (ISGylation). Several target proteins for ISGylation have recently been identified, but the biological consequence of protein ISGylation remains unclear. In the course of our study to identify components of the ISGylation system, we found that Ubc13, an E2 enzyme for ubiquitin conjugation, is covalently modified with ISG15. To determine the meaning of ISGylation of Ubc13, we isolated ISG15-modified Ubc13 protein and compared its ubiquitin-conjugating activity with that of an unmodified one. We found that ISGylation of Ubc13 suppresses its ability to form a thioester intermediate with ubiquitin.