EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Salmonella enterica serotype Typhimurium fimbrial proteins serve as antigens during infection of mice.
PMID 16113248
Abstract
The Salmonella enterica serotype Typhimurium genome contains 13 operons with homology to fimbrial gene sequences. Here we investigated the role of 11 serotype Typhimurium fimbrial proteins, including FimA, AgfA (CsgA), BcfA, StbA, SthA, LpfA, PefA, StdA, StcA, StiA, and StfA, as antigens during the infection of genetically resistant mice (CBA). Upon the growth of serotype Typhimurium in standard laboratory broth culture, only the expression of FimA could be detected by Western blot analysis. The infection of mice with serotype Typhimurium grown in broth culture, followed by at least one subsequent infection, resulted in seroconversion of animals to FimA, AgfA, BcfA, StbA, SthA, LpfA, PefA, StdA, StcA, StiA, and StfA positivity. Most animals seroconverted to only a subset of these fimbrial antigens. The immunization of mice with glutathione S-transferase (GST)-FimA, GST-AgfA, GST-BcfA, GST-StbA, GST-SthA, GST-LpfA, GST-PefA, GST-StdA, GST-StcA, GST-StiA, and GST-StfA fusion proteins resulted in reduced fecal shedding of serotype Typhimurium during a challenge compared to that by a control group immunized with purified GST protein. Collectively, these data suggest that the expression of serotype Typhimurium fimbrial antigens is induced during the infection of mice.