EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Acquired predisposition to mycobacterial disease due to autoantibodies to IFN-gamma.
PMID 16127458
Abstract
Genetic defects in the IFN-gamma response pathway cause unique susceptibility to intracellular pathogens, particularly mycobacteria, but are rare and do not explain mycobacterial disease in the majority of affected patients. We postulated that acquired defects in macrophage activation by IFN-gamma may cause a similar immunological phenotype and thus explain the occurrence of disseminated intracellular infections in some patients without identifiable immune deficiency. Macrophage activation in response to IFN-gamma and IFN-gamma production were studied in whole blood and PBMCs of 3 patients with severe, unexplained nontuberculous mycobacterial infection. In all 3 patients, IFN-gamma was undetectable following mitogen stimulation of whole blood, but significant quantities were detectable in the supernatants of PBMCs when stimulated in the absence of the patients' own plasma. The patients' plasma inhibited the ability of IFN-gamma to increase production of TNF-alpha by both autologous and normal donor PBMCs, and recovery of exogenous IFN-gamma from the patients' plasma was greatly reduced. Using affinity chromatography, surface-enhanced laser desorption/ionization mass spectrometry, and sequencing, we isolated an IFN-gamma-neutralizing factor from the patients' plasma and showed it to be an autoantibody against IFN-gamma. The purified anti-IFN-gamma antibody was shown to be functional first in blocking the upregulation of TNF-alpha production in response to endotoxin; second in blocking induction of IFN-gamma-inducible genes (according to results of high-density cDNA microarrays); and third in inhibiting upregulation of HLA class II expression on PBMCs. Acquired defects in the IFN-gamma pathway may explain unusual susceptibility to intracellular pathogens in other patients without underlying, genetically determined immunological defects.