EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
A small compound that inhibits tumor necrosis factor-alpha-induced matrix metalloproteinase-9 upregulation.
PMID 16140265
Abstract
Matrix metalloproteinase-9 (MMP-9) is critically involved in the tumor invasion and metastasis processes. Since TNF-alpha plays a crucial role in the regulation of MMP-9 expression, the development of molecules capable of modulating TNF-alpha-induced signaling is an issue of concern. We identified a novel synthetic compound that inhibits TNF-alpha-induced MMP-9 upregulation in the HT1080 human fibrosarcoma cell line. The active compound SM-7368 inhibited TNF-alpha-induced MMP-9 upregulation in a concentration-dependent manner and showed maximal activity at 10 microM. SM-7368 inhibited TNF-alpha-induced MMP-9 mRNA transcript accumulation and protein expression. We also found that SM-7368 strongly inhibits TNF-alpha-induced NF-kappaB activity but not AP-1 activity. Moreover, we found that SM-7368 strongly inhibits the TNF-alpha-induced invasion of HT1080 human fibrosarcoma cell line. Taken together, our results demonstrate that SM-7368 is a synthetic compound that inhibits TNF-alpha-induced MMP-9 expression, and thus SM-7368 should be useful for the development of chemotherapies targeting TNF-alpha-mediated tumor invasion and metastasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0454
SM-7368, ≥98% (HPLC)
C10H5ClN4O5S