EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Reactions of D-glucose with phenolic amino acids: further insights into the competition between Maillard and Pictet-Spengler condensation pathways.
PMID 16229826
Abstract
The reactions of 5-S-cysteinyldopa, L-alpha-methyldopa and DL-m-tyrosine with D-glucose were investigated at 90 degrees C in phosphate buffer at pH ranging from 5.0 to 9.0. Whereas gave mainly the double Maillard condensation product N,N'-bis(1''-deoxy-D-fructos-1''-yl)-5-S-cysteinyldopa, as an inseparable mixture of beta-D-fructopyranosyl and alpha,beta-D-fructofuranosyl derivatives, 2 and 3 gave both Maillard and Pictet-Spengler products, although to different extents and with different regio- and stereochemistry. A peculiar pattern of reactivity was displayed by which gave, besides the Maillard product and the expected 6,7-dihydroxy-1,2,3,4-tetrahydroisoquinoline C-1 diastereoisomeric pairs, the unprecedented 7,8-dihydroxy-1,2,3,4-tetrahydroisoquinoline derivative via the ortho cyclization pathway. Pictet-Spengler cyclization of 2 and 3 proceeded with Felkin-Anh-type asymmetric induction, favouring the 1R isomer throughout the pH range 5.0-9.0. These results, which highlight the first example of carbohydrate-derived 7,8-dihydroxytetrahydroisoquinoline, provide new insights into the factors governing competition between Maillard and Pictet-Spengler condensation pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3629
DL-m-Tyrosine, crystalline
C9H11NO3