EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Nitrosubstituted aromatic compounds as nitrogen source for bacteria.
PMID 16347259
Abstract
Bacteria which utilized nitroaromatic compounds (0.5 mM) as sole source of nitrogen were isolated from soil. With 2,6-dinitrophenol and succinate as carbon source, a Pseudomonas strain was isolated which liberated and assimilated nitrite. Approximately 2 mol of NO(2) per mol of 2,6-dinitrophenol was released by resting cells. The xenobiotic compound was totally degraded, although specific growth yields were low even with succinate as a carbon source.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42155
2,3-Dinitrophenol, moistened with water, ≥98.0% (HPLC)
C6H4N2O5