EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Dammarane triterpenes from the hypocotyls and fruits of Ceriops tagal.
PMID 16378377
Abstract
Three new dammarane triterpenes, cereotagaloperoxide, cereotagalol A, and cereotagalol B, together with four known dammarane triterpenes, an oleanane triterpene, and 13 known lupane triterpenes were isolated from the hypocotyls and fruits of Ceriops tagal. The structures of 1-3 were characterized on the basis of their spectroscopic data.