EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Rapid sequencing and disulfide mapping of peptides containing disulfide bonds by using 1,5-diaminonaphthalene as a reductive matrix.
PMID 16382486
Abstract
MS/MS is indispensable for the amino acid sequencing of peptides. However, its use is limited for peptides containing disulfide bonds. We have applied the reducing properties of 1,5-diaminonaphthalene (1,5-DAN) as a MALDI matrix to amino acid sequencing and disulfide bond mapping of human urotensin II possessing one disulfide bond, and human guanylin possessing two disulfide bonds. 1,5-DAN was used in the same manner as the usual MALDI matrices without any pre-treatment of the peptide, and MS/MS was performed using a matrix-assisted laser desorption/ionization quadrupole ion trap time-of-flight mass spectrometer (MALDI QIT TOFMS). The results demonstrated that MS/MS of the molecular ions reduced by 1,5-DAN provided a series of significant b-/y-product ions. All 11 amino acid residues of urotensin II were identified using 1,5-DAN, while only 5 out of 11 residues were identified using 2,5-dihydroxybenzoic acid (DHB); similarly 11 out of 15 amino acid residues of guanylin were identified using 1,5-DAN, while only three were identified using DHB. In addition, comparison of the theoretical and measured values of the mass differences between corresponding MS/MS product ions using 1,5-DAN and DHB narrowed down the possible disulfide bond arrangement candidates. Consequently, 1,5-DAN as a reductive matrix facilitates rapid amino acid sequencing and disulfide mapping for peptides containing disulfide bonds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56451
1,5-Diaminonaphthalene, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C10H10N2