EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Substituted 2-thioxoimidazolidin-4-ones and imidazolidine-2,4-diones as fatty acid amide hydrolase inhibitors templates.
PMID 16392827
Abstract
The demonstration of the essential role of fatty acid amide hydrolase (FAAH) in hydrolyzing endogenous bioactive fatty acid derivatives has launched the quest for the discovery of inhibitors for this enzyme. Along this line, a set of 58 imidazolidine-2,4-dione and 2-thioxoimidazolidin-4-one derivatives was evaluated as FAAH inhibitors. Among these compounds, 3-substituted 5,5'-diphenylimidazolidine-2,4-dione and 3-substituted 5,5'-diphenyl-2-thioxoimidazolidin-4-one derivatives were previously described as CB(1) cannabinoid receptor ligands. In the present study, we synthesized several derivatives exhibiting interesting FAAH inhibitory activity and devoid of affinity for the CB(1) and CB(2) cannabinoid receptors. For instance, 3-heptyl-5,5'-diphenylimidazolidine-2,4-dione (14) and 5,5'-diphenyl-3-tetradecyl-2-thioxo-imidazolidin-4-one (46) showed pI(50) values of 5.12 and 5.94, respectively. In conclusion, it appears that even though several 3-substituted 5,5'-diphenyl-2-thioxoimidazolidin-4-one and 3-substituted 5,5'-diphenylimidazolidine-2,4-dione derivatives have been previously shown to behave as CB(1) cannabinoid receptor ligands, appropriate substitutions of these templates can result in FAAH inhibitors devoid of affinity for the cannabinoid receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D214256
5,5-Diphenyl-2-thiohydantoin, 99%
C15H12N2OS