EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Bacterial biofilm inhibitors from Diospyros dendo.
PMID 16441080
Abstract
One new (1) and four known (2-5) ursene triterpenes with potent inhibition of the formation of the bacterial biofilm Pseudomonas aeruginosa PA01 were obtained from Diospyros dendo using a high-throughput natural products chemistry procedure. These compounds were isolated as mass-limited samples. The miniaturization of the structure elucidation and dereplication was performed primarily utilizing a capillary-scale NMR probe.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89797
Ursolic acid, analytical standard
C30H48O3