EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Low molecular weight indole fragments as IMPDH inhibitors.
PMID 16483769
Abstract
The study of non-oxazole containing indole fragments as inhibitors of inosine monophosphate dehydrogenase (IMPDH) is described. The synthesis and in vitro inhibitory values for IMPDH II are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
347949
3-Cyanoindole, 98%
C9H6N2
638854
7-Nitroindole, 97%
C8H6N2O2
129445
Indole-3-carboxaldehyde, 97%
C9H7NO