EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
3-[6-(2-Dimethylamino-1-imidazol-1-yl-butyl)-naphthalen-2-yloxy]-2,2-dimethyl-propionic acid as a highly potent and selective retinoic acid metabolic blocking agent.
PMID 16504501
Abstract
3-[6-(2-Dimethylamino-1-imidazol-1-yl-butyl)-naphthalen-2-yloxy]-2,2-dimethyl-propionic acid and analogs were designed and synthesized as highly potent and selective CYP26 inhibitors, serving as retinoic acid metabolic blocking agents (RAMBAs), with demonstrated in vivo efficacy to increase the half-life of exogenous atRA.