EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Catalytic, efficient, and syn-selective construction of deoxypolypropionates and other chiral compounds via Zr-catalyzed asymmetric carboalumination of allyl alcohol.
PMID 16506734
Abstract
An efficient, syn-selective, all catalytic asymmetric protocol for the synthesis of alpha,omega-diheterofunctional deoxypolypropionates via Zr-catalyzed asymmetric carboalumination (ZACA reaction) was developed. The success of the method critically hinges on the one-pot conversion of unprotected allyl alcohol into TBS-protected (R)- or (S)-3-iodo-2-methyl-1-propanol (1) of 91% enantiomeric purity in 82% yield via (i) Zr-catalyzed asymmetric methylalumination, (ii) iodination, and (iii) protection with TBSCl. After zincation of 1, its Pd-catalyzed cross-coupling with various organic halides can give various organic derivatives, including 3 and 4, which can serve as key intermediates for efficient and selective syntheses of deoxypolypropionates, such as doliculide and 2,4,6,8-tetramethyldecanoic acid, and other chiral compounds, such as callystatin A. Selective monocarboalumination of 1,4-pentadiene (5 equiv) gave, after oxidation, the expected product 5, but it was 0% ee. A plausible mechanism for racemization has been proposed and experimentally supported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
719072
Dichlorobis(1-neomenthylindenyl)-zirconium, 95%
C38H50Cl2Zr