EMAIL THIS PAGE TO A FRIEND
Biopolymers
2-(N-Fmoc)-3-(N-Boc-N-methoxy)-diaminopropanoic acid, an amino acid for the synthesis of mimics of O-linked glycopeptides.
PMID 16508952
Abstract
Amino acids with N-alkylaminooxy side chains have proven effective for the rapid synthesis of neoglycopeptides. Chemoselective reaction of reducing sugars with peptides containing these amino acids provides glycoconjugates that are structurally similar to their natural counterparts. 2-(N-Fmoc)-3-(N-Boc-N-methoxy)-diaminopropanoic acid (Fmoc: 9-fluorenylmethoxycarbonyl; Boc: t-butyloxycarbonyl) was synthesized from Boc-Ser-OH in >40% overall yield and incorporated into peptides by standard Fmoc chemistry based solid phase peptide synthesis. The resulting peptides are efficiently glycosylated and serve as mimics of O-linked glycopeptides. The synthesis of this derivative greatly expands the availability of the N-alkylaminooxy strategy for neoglycopeptides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15500
Boc-Ser-OH, ≥99.0% (T)
C8H15NO5