EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
Genomic and expression analysis of the 3q25-q26 amplification unit reveals TLOC1/SEC62 as a probable target gene in prostate cancer.
PMID 16547154
Abstract
Gain at chromosome 3q25-q26 has been reported to commonly occur in prostate cancer. To map the 3q25-q26 amplification unit and to identify the candidate genes of amplification, we did fluorescence in situ hybridization and quantitative real-time PCR for gene copy number and mRNA expression measurements in prostate cancer cell lines and prostate cancer samples from radical prostatectomy specimens. The minimal overlapping region of DNA copy number gains in the cell lines could be narrowed down to 700 kb at 3q26.2. Of all positional and functional candidates in this region, the gene TLOC1/SEC62 revealed the highest frequency (50%) of copy number gains in the prostate cancer samples and was found to be up-regulated at the mRNA level in all samples analyzed. TLOC1/Sec62 protein was also shown to be overexpressed by Western blot analysis. Intriguingly, the TLOC1/SEC62 gene copy number was increased in prostate tumors from patients who had a lower risk of and a longer time to progression following radical prostatectomy. These findings make TLOC1/SEC62 the best candidate within the 3q amplification unit in prostate cancer. TLOC1/Sec62 protein is a component of the endoplasmic reticulum protein translocation machinery, whose function during prostate carcinogenesis remains to be determined.