EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
The Inhibition of the Spreading Growth of Proteus and other Bacteria to Permit the Isolation of Associated Streptococci.
PMID 16560477
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70132 Azide Blood Agar (Base), for microbiology